CAS RN 65373-29-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ MS 21570
GPR171 antagonist
2
MS21570, MedChemExpress
MedChemExpress MS21570 is a selective GPR171 antagonist, with an IC50 of 220 nM.