CAS RN 64585-12-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Ethyl-2,2,2-d3-amine HCl, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Ethylamine Hydrochloride-d3, TRC
CAS: 64585-12-2 Molekylær formel: C2 D3 H4 N . Cl H Molekylvægt (g/mol): 84.563 Synonym: Ethan-2,2,2-d3-amine, hydrochloride (9CI),2,2,2-Trideuterioethanamine hydrochloride,Ethan-2,2,2-d3-amine hydrochloride,Ethylamine-d3 hydrochloride,N-(Ethyl-2,2,2-d3)ammonium chloride,Ethylamine-2,2,2-D3 hydrochloride IUPAC navn: 2,2,2-trideuterioethanamine;hydrochloride SMIL: Cl.[2H]C([2H])([2H])CN