Få mere at vide
Ethylamine Hydrochloride-d3, TRC
Artikelnummer. 30180384
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180384 5 mg
30179884 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180384 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8982935MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64585-12-2
Molekylær formel C2 D3 H4 N . Cl H
Molekylvægt (g/mol) 84.563
Synonym Ethan-2,2,2-d3-amine, hydrochloride (9CI), 2,2,2-Trideuterioethanamine hydrochloride, Ethan-2,2,2-d3-amine hydrochloride, Ethylamine-d3 hydrochloride, N-(Ethyl-2,2,2-d3)ammonium chloride, Ethylamine-2,2,2-D3 hydrochloride
IUPAC navn 2,2,2-trideuterioethanamine;hydrochloride
SMIL Cl.[2H]C([2H])([2H])CN
Tekniske data
Kemisk navn eller materiale Ethylamine Hydrochloride-d3
Mængde 5 mg
InChI formel InChI=1S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1H/i1D3;
Formel vægt 84.053