CAS RN 64284-64-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(3beta,5alpha)-4,4-Dimethylcholesta-8,14,24-trien-3-ol(Contain up to 15% ∆25 isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.