CAS RN 638-66-4
IUPAC Navn:
octadecanal
Synonymer:
octadecanal
Molekylvægt (g/mol):
268.49
Molekylformel:
C18H36O
InChi Key:
FWWQKRXKHIRPJY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC=O
1
–
2
af
2
Resultater
1
Octadecanaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Octadecanaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.