CAS RN 634-90-2
IUPAC Navn:
1,2,3,5-tetrachlorobenzene
Synonymer:
1,2,3,5-tetrachlorobenzene C6H2cl4
Molekylvægt (g/mol):
215.88
Molekylformel:
C6H2Cl4
InChi Key:
QZYNWJQFTJXIRN-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(Cl)C(Cl)=C1
1
–
6
af
6
Resultater
2
1,2,3,5-Tetrachlorobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3,5-Tetrachlorobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.