CAS RN 608-73-1
IUPAC Navn:
1,2,3,4,5,6-hexachlorocyclohexane
Synonymer:
lindane hexachlorocyclohexane 林旦 hexachlorocyclohexanes hexachlorzyklohexan hexachlorcyclohexan benzene hexachloride
Molekylvægt (g/mol):
290.81
Molekylformel:
C6H6Cl6
InChi Key:
JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES:
ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
1
–
4
af
4
Resultater
1
BHC, mixture of isomers (a:b:gamma:delta=1:1:1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
BHC, mixture of isomers (Alpha:Beta:Gamma:Delta=1:1:1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
BHC, mixture of isomers (a:b:gamma:delta=1:1:1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.