CAS RN 608-21-9
IUPAC Navn:
1,2,3-tribromobenzene
Synonymer:
1,2,3-tribromobenzene
Molekylvægt (g/mol):
314.8
Molekylformel:
C6H3Br3
InChi Key:
GMVJKSNPLYBFSO-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(Br)=C1Br
1
–
3
af
3
Resultater
1
1,2,3-Tribromobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3-Tribromobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3-Tribromobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.