CAS RN 607737-87-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ A 77-01
Potent inhibitor of TGF-βRI
2
A 77-01, MedChemExpress
MedChemExpress A 77-01 is a potent inhibitor of transforming growth factor (TGF)-β type I receptor superfamily activin-like kinase ALK5 with an IC50 of 25 nM.