Få mere at vide
A 77-01, MedChemExpress
Artikelnummer. 16460678
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16460678 5 mg
16428238 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16460678 Leverandør Medchem Express Leverandørnr. HY78349
MedChemExpress A 77-01 is a potent inhibitor of transforming growth factor (TGF)-β type I receptor superfamily activin-like kinase ALK5 with an IC50 of 25 nM.
A 77-01 is a potent inhibitor of transforming growth factor (TGF)-β type I receptor superfamily activin-like kinase ALK5 with an IC50 of 25 nM.
Tekniske data
Kemisk navn eller materiale A 77-01
CAS 607737-87-1
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C18H14N4
Mængde 5 mg
Opløselighedsinformation DMSO : 25 mg/mL (87.31 mM; Need ultrasonic)
SMIL CC1=CC=CC(C2=NNC=C2C3=CC=NC4=CC=CC=C34)=N1
Molekylvægt (g/mol) 286.33
Formel vægt 286.33
Procent renhed 99.55%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease