Få mere at vide
Tocris Bioscience™ A 77-01
Beskrivelse
A 77-01 is a potent TGF-βRI (ALK-5) inhibitor (IC50 = 34 nM). Inhibits TGF-β transcriptional activation. Inhibits TGF-β-induced proliferation of lung epithelial cells and SMAD phosphorylation. Inhibits TGF-β-induced epithelial-to-mesenchymal transition (EMT). Also inhibitor of p38 MAPK. Likely active metabolite of A 83-01 (Cat. No. 2939).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[3-(6-Methyl-2-pyridinyl)-1H-pyrazol-4-yl]quinoline |
| CAS | 607737-87-1 |
| Mængde | 5 mg |
| Molekylær formel | C18H14N4 |
| Formel vægt | 286.33 |
| Renhed | 98% |