CAS RN 6055-52-3
IUPAC Navn:
hexane-1,6-bis(aminium) dichloride
Synonymer:
hexane-1,6-bis(aminium) dichloride
Molekylvægt (g/mol):
189.12
Molekylformel:
C6H18Cl2N2
InChi Key:
XMVQMBLTFKAIOX-UHFFFAOYSA-N
SMILES:
[Cl-].[Cl-].[NH3+]CCCCCC[NH3+]
1
–
2
af
2
Resultater
1
1,6-Diaminohexane Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hexane-1,6-diamine Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.