CAS RN 60-57-1
IUPAC Navn:
3,4,5,6,13,13-hexachloro-10-oxapentacyclo[6.3.1.1³,⁶.0²,⁷.0⁹,¹¹]tridec-4-ene
Synonymer:
endrin dieldrin (iso)
Molekylvægt (g/mol):
380.9
Molekylformel:
C12H8Cl6O
InChi Key:
DFBKLUNHFCTMDC-UHFFFAOYNA-N
SMILES:
ClC1=C(Cl)C2(Cl)C3C4CC(C5OC45)C3C1(Cl)C2(Cl)Cl
1
–
9
af
9
Resultater
3
Dieldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Dieldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Dieldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.