CAS RN 59886-90-7
IUPAC Navn:
4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Synonymer:
4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Molekylvægt (g/mol):
158.61
Molekylformel:
C7H9ClNO
InChi Key:
HHYDNQMGLBHVSW-UHFFFAOYNA-N
SMILES:
CC1=C(C)C(Cl)=C[CH-][NH++]1[O-]
1
–
2
af
2
Resultater
1
4-Chloro-2,3-dimethylpyridine N-Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Chloro-2,3-dimethylpyridine N-Oxide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.