Få mere at vide
4-Chloro-2,3-dimethylpyridine N-Oxide, Mikromol™
Artikelnummer. 30462518
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462518 4 x 25 mg
30049424 25 mg
17952526 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462518 Leverandør Mikromol Leverandørnr. MM0618.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Chloro-2,3-dimethylpyridine N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-2,3-dimethylpyridine N-Oxide
CAS 59886-90-7
Analyt- eller komponentnavne 4-Chloro-2,3-dimethylpyridine N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C7 H8 Cl N O
Certificeringer/compliance ISO 9001
Synonym 4-Chloro-2,3-dimethylpyridine N-Oxide
InChI formel InChI=1S/C7H8ClNO/c1-5-6(2)9(10)4-3-7(5)8/h3-4H,1-2H3
Løsningstype Neat
SMIL Cc1c(C)[n+]([O-])ccc1Cl
IUPAC navn 4-chloro-2,3-dimethyl-1-oxidopyridin-1-ium
Molekylvægt (g/mol) 157.6
Mængde 4 x 25 mg
Formel vægt 157.0294 g/mol