CAS RN 5980-31-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2,6-Bis(2-ethylhexyl)-7a-methylhexahydro-1H-imidazo[1,5-c]imidazole (Hexedine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Hexedine, TRC
CAS: 5980-31-4 Molekylær formel: C22 H45 N3 Molekylvægt (g/mol): 351.61 Synonym: 2,6-Bis(2-ethylhexyl)-7a-methylhexahydro-1H-imidazo[1,5-c]imidazole,Hexetidine Imp. C (EP),Hexedine IUPAC navn: 2,6-bis(2-ethylhexyl)-7a-methyl-1,3,5,7-tetrahydroimidazo[1,5-c]imidazole SMIL: CCCCC(CC)CN1CN2CN(CC(CC)CCCC)CC2(C)C1