CAS RN 59469-74-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(6RS)-8-Chloro-6-(2-fluorophenyl)-1-methyl-6H-imidazo[1,5-a][1,4]benzodiazepine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
8-Chloro-6-(2-fluorophenyl)-1-methyl-6H-Imidazo[1,5-a][1,4]benzodiazepine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
8-Chloro-6-(2-fluorophenyl)-1-methyl-6H-Imidazo[1,5-a][1,4]benzodiazepine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.