CAS RN 590376-39-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-Allyl-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Allyl-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Allyl-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.