missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Allyl-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol, TRC

Artikelnummer. 30311788
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30311788 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311788 Supplier Toronto Research Chemicals Supplier No. TRCA614753500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Allyl-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol
CAS 590376-39-9
Molekylær formel C13H15N3S
Mængde 500 mg
Synonym 2,4-Dihydro-5-(2-phenylethyl)-4-(2-propen-1-yl)-3H-1,2,4-triazole-3-thione
InChI formel InChI=1S/C13H15N3S/c1-2-10-16-12(14-15-13(16)17)9-8-11-6-4-3-5-7-11/h2-7H,1,8-10H2,(H,15,17)
SMIL C=CCN1C(=S)NN=C1CCc2ccccc2
IUPAC navn 3-(2-phenylethyl)-4-prop-2-enyl-1H-1,2,4-triazole-5-thione
Molekylvægt (g/mol) 245.34
Formel vægt 245.0987
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.