CAS RN 587841-73-4
IUPAC Navn:
1-[2-(4-bromo-2-chlorophenoxy)acetyl]-3-{4-[(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl}thiourea
Synonymer:
1-[2-(4-bromo-2-chlorophenoxy)acetyl]-3-{4-[(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl}thiourea
Molekylvægt (g/mol):
584.89
Molekylformel:
C21H19BrClN5O4S2
InChi Key:
XKZDWYDHEBCGCG-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=NC(NS(=O)(=O)C2=CC=C(NC(=S)NC(=O)COC3=C(Cl)C=C(Br)C=C3)C=C2)=N1
1
–
1
af
1
Resultater
1
ZCL278, MedChemExpress
MedChemExpress ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with Kd of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment.