CAS RN 574013-66-4
IUPAC Navn:
methyl (2E)-3-[3-(N-{[4'-(dimethylamino)-[1,1'-biphenyl]-4-yl]methyl}cyclohexaneamido)phenyl]prop-2-enoate
Synonymer:
fexaramine
Molekylvægt (g/mol):
496.65
Molekylformel:
C32H36N2O3
InChi Key:
VLQTUNDJHLEFEQ-KGENOOAVSA-N
SMILES:
COC(=O)\C=C\C1=CC=CC(=C1)N(CC1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C)C(=O)C1CCCCC1
1
–
2
af
2
Resultater
1
Tocris Bioscience™ Fexaramine
CAS: 574013-66-4 Molekylær formel: C32H36N2O3
2
Fexaramine, MedChemExpress
MedChemExpress Fexaramine is a potent and selective FXR agonist with an EC50 of 25 nM. Fexaramine has no activity against hRXRα, hPPARαγδ, mPXR, hPXR, hLXRα, hTRβ, hRARβ, mCAR, mERRγ, and hVDR receptors.