CAS RN 57149-07-2
IUPAC Navn:
1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-(naphthalen-1-yloxy)propan-2-ol
Synonymer:
naftopidil flivas naftopidilum
Molekylvægt (g/mol):
392.5
Molekylformel:
C24H28N2O3
InChi Key:
HRRBJVNMSRJFHQ-UHFFFAOYNA-N
SMILES:
COC1=CC=CC=C1N1CCN(CC(O)COC2=C3C=CC=CC3=CC=C2)CC1
1
–
2
af
2
Resultater
1
Naftopidil, TRC
CAS: 57149-07-2 Molekylær formel: C24 H28 N2 O3 Molekylvægt (g/mol): 392.49 IUPAC navn: 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol SMIL: COc1ccccc1N2CCN(CC(O)COc3cccc4ccccc34)CC2
2
Naftopidil, MedChemExpress
MedChemExpress Naftopidil (KT-611) is is a selective alpha1-adrenoceptor antagonist, with Kis of 3.7 nM, 20 nM and 1.2 nM for the cloned human α1a-, α1b- and α1d-adrenoceptor subtypes, respectively. Naftopidil has antiproliferative effects. Naftopidil can be used for the research of prostate hyperplasia.