Få mere at vide
Naftopidil, TRC
Artikelnummer. 30480871
Skift visning
Mængde:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480871 25 mg
30466436 500 mg
30471528 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480871 Leverandør Toronto Research Chemicals Leverandørnr. TRCN21350125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57149-07-2
Molekylær formel C24 H28 N2 O3
Molekylvægt (g/mol) 392.49
IUPAC navn 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol
SMIL COc1ccccc1N2CCN(CC(O)COc3cccc4ccccc34)CC2
Tekniske data
Kemisk navn eller materiale Naftopidil
Mængde 25 mg
InChI formel InChI=1S/C24H28N2O3/c1-28-24-11-5-4-10-22(24)26-15-13-25(14-16-26)17-20(27)18-29-23-12-6-8-19-7-2-3-9-21(19)23/h2-12,20,27H,13-18H2,1H3
Formel vægt 392.21