CAS RN 5627-05-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
5,6-Dihydrouridine, TRC
CAS: 5627-05-4 Molekylær formel: C9H14N2O6 Molekylvægt (g/mol): 246.22 Synonym: Dihydro-1-β-D-ribofuranosyl-2,4(1H,3H)-pyrimidinedione,Dihydrouridine,1-β-D-Ribofuranosyl-hydrouracil; SMIL: O=C(NC1=O)CCN1[C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2
2
5,6-Dihydrouridine, MedChemExpress
MedChemExpress 5,6-Dihydrouridine is a modified base found in conserved positions in the D-loop of tRNA in Bacteria, Eukaryota, and some Archaea.