CAS RN 541-91-3
IUPAC Navn:
3-methylcyclopentadecan-1-one
Synonymer:
methylcyclopentadecanone
Molekylvægt (g/mol):
238.42
Molekylformel:
C16H30O
InChi Key:
ALHUZKCOMYUFRB-UHFFFAOYNA-N
SMILES:
CC1CCCCCCCCCCCCC(=O)C1
1
–
3
af
3
Resultater
1
Muscone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Muscone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.