Få mere at vide
Muscone, TRC
Artikelnummer. 30482044
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482044 500 mg
30468607 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482044 Leverandør Toronto Research Chemicals Leverandørnr. TRCM815090500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methylcyclopentadecanone
CAS 541-91-3
Molekylær formel C16 H30 O
Mængde 500 mg
Synonym Muscone (6CI), 3-Methylcyclopentadecanone, (±)-Muscone, 3-Methylcyclopentadecan-1-one, Moschus ketone, Muskone, dl-Muscone, Cyclopentadecanone','3-methyl-
InChI formel InChI=1S/C16H30O/c1-15-12-10-8-6-4-2-3-5-7-9-11-13-16(17)14-15/h15H,2-14H2,1H3
SMIL CC1CCCCCCCCCCCCC(=O)C1
IUPAC navn 3-methylcyclopentadecan-1-one
Molekylvægt (g/mol) 238.41
Formel vægt 238.2297