CAS RN 53907-55-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Isonicotinic-d4 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Isonicotinic Acid-d4, TRC
CAS: 53907-55-4 Molekylær formel: C6 D4 H N O2 Molekylvægt (g/mol): 127.1341 Synonym: 4-Pyridine-2,3,5,6-d4-carboxylic acid (9CI),Isonicotinic-d4 Acid,Isonicotinic-D4 Acid,Pyridine-4-carboxylic acid-d4,Isonicotinic acid-d4 IUPAC navn: 2,3,5,6-tetradeuteriopyridine-4-carboxylic acid SMIL: [2H]c1nc([2H])c([2H])c(C(=O)O)c1[2H]