Få mere at vide
Isonicotinic Acid-d4, TRC
Artikelnummer. 30180127
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180127 250 mg
30180111 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180127 Leverandør Toronto Research Chemicals Leverandørnr. TRCI821762250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53907-55-4
Molekylær formel C6 D4 H N O2
Molekylvægt (g/mol) 127.1341
Synonym 4-Pyridine-2,3,5,6-d4-carboxylic acid (9CI), Isonicotinic-d4 Acid, Isonicotinic-D4 Acid, Pyridine-4-carboxylic acid-d4, Isonicotinic acid-d4
IUPAC navn 2,3,5,6-tetradeuteriopyridine-4-carboxylic acid
SMIL [2H]c1nc([2H])c([2H])c(C(=O)O)c1[2H]
Tekniske data
Kemisk navn eller materiale Isonicotinic Acid-d4
Mængde 250 mg
InChI formel InChI=1S/C6H5NO2/c8-6(9)5-1-3-7-4-2-5/h1-4H,(H,8,9)/i1D,2D,3D,4D
Formel vægt 127.0571