Få mere at vide
Isonicotinic-d4 Acid, CDN
Artikelnummer. 30319818
Skift visning
Mængde:
0.1 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319818 0.1 g -
30316980 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319818 Leverandør CDN Leverandørnr. CDND43480.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isonicotinic Acid-d4
CAS 53907-55-4
Molekylær formel C6 D4 H N O2
Mængde 0.1 g
Synonym 4-Pyridine-2,3,5,6-d4-carboxylic acid (9 CI), Isonicotinic-d4 Acid, Isonicotinic-D4 Acid, Pyridine-4-carboxylic acid-d4, Isonicotinic acid-d4
InChI formel InChI=1 S/C6H5NO2/c8-6(9)5-1-3-7-4-2-5/h1-4 H,(H,8,9)/i1D,2 D,3 D,4 D
SMIL [2 H]c1nc([2 H])c([2 H])c(C(=O)O)c1[2 H]
IUPAC navn 2,3,5,6-tetradeuteriopyridine-4-carboxylic acid
Molekylvægt (g/mol) 127.1341
Formel vægt 127.0571 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity