CAS RN 536-25-4
IUPAC Navn:
methyl 3-amino-4-hydroxybenzoate
Synonymer:
orthoform orthoderm orthocaine 奥索卡因
Molekylvægt (g/mol):
167.16
Molekylformel:
C8H9NO3
InChi Key:
VNQABZCSYCTZMS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(O)C(N)=C1
1
–
3
af
3
Resultater
1
Orthocaine, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Orthocaine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Orthocaine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.