Få mere at vide
Orthocaine, LoGiCal
Artikelnummer. 30462376
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462376 10 mg
1 options
Artikelnummer. 30462376 Leverandør LoGiCal Leverandørnr. LGCFOR3166.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Orthocaine
CAS 536-25-4
Analyt- eller komponentnavne Orthocaine
Forsendelsestilstand Room Temperature
Molekylær formel C8 H9 N O3
Certificeringer/compliance ISO 9001
Synonym Benzoic acid, 3-amino-4-hydroxy-, methyl ester, Orthoform (6CI), 1-Hydroxy-2-amino-4-carbomethoxybenzene, 3-Amino-4-hydroxybenzoic acid methyl ester, Aminobenz, Methyl 3-amino-4-hydroxybenzoate, Methyl 4-hydroxy-3-aminobenzoate, Methyl m-amino-p-hydroxybenzoate, NSC 9813, Orthocaine, Orthoderm, Orthoform New
InChI formel InChI=1S/C8H9NO3/c1-12-8(11)5-2-3-7(10)6(9)4-5/h2-4,10H,9H2,1H3
Løsningstype Neat
SMIL COC(=O)c1ccc(O)c(N)c1
IUPAC navn methyl 3-amino-4-hydroxybenzoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 167.16
Mængde 10 mg
Formel vægt 167.0582 g/mol