CAS RN 5341-49-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(5RS)-6-(Dimethylamino)-5-methyl-4,4-diphenylhexan-3-one Hydrochloride (Isomethadone Hydrochloride), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.