Få mere at vide
(5RS)-6-(Dimethylamino)-5-methyl-4,4-diphenylhexan-3-one Hydrochloride (Isomethadone Hydrochloride), Mikromol
Beskrivelse
(5RS)-6-(Dimethylamino)-5-methyl-4,4-diphenylhexan-3-one Hydrochloride (Isomethadone Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isomethadone Hydrochloride |
| CAS | 5341-49-1 |
| Analyt- eller komponentnavne | Isomethadone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H27 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Isomethadone hydrochloride, NSC 2106, 3-Hexanone, 6-(dimethylamino)-5-methyl-4,4-diphenyl-, hydrochloride (7CI,8CI,9CI), Imp. D (EP) as Hydrochloride: (5RS)-6-(Dimethylamino)-5-methyl-4,4-diphenylhexan-3-one Hydrochloride (Isomethadone Hydrochloride), Methadone Related Compound D as Hydrochloride, Methadone Hydrochloride Impurity D as Hydrochloride, Methadone Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C21H27NO.ClH/c1-5-20(23)21(17(2)16-22(3)4,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6-15,17H,5,16H2,1-4H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCC(=O)C(C(C)CN(C)C)(c1ccccc1)c2ccccc2 |
| Vis mere |