CAS RN 53001-22-2
CAS RN 53001-22-2
IUPAC Navn:
2-(²H₃)methyl(²H₆)propan-2-(²H)ol
Synonymer:
2-(²H₃)methyl(²H₆)propan-2-(²H)ol
Molekylvægt (g/mol):
84.18
Molekylformel:
C4H10O
InChi Key:
DKGAVHZHDRPRBM-SGLLWXCUSA-N
SMILES:
[2H]OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
1
–
4
af
4
Resultater
1
tert-Butyl Alcohol-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl Alcohol-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
tert-Butyl Alcohol-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
tert-Butyl Alcohol-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.