Få mere at vide
tert-Butyl Alcohol-d10, TRC
Artikelnummer. 30418776
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30418776 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418776 Leverandør Toronto Research Chemicals Leverandørnr. TRCT117541250MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butyl Alcohol-d10
CAS 53001-22-2
Molekylær formel C4 D10 O
Mængde 250 mg
Synonym 2-Propan-1,1,1,3,3,3-d6-ol-d','2-(methyl-d3)- (9CI), tert-Butyl-d9 alcohol-d (6CI','7CI), 1,1,1,3,3,3-Hexadeutero-2-deuterooxy-2-(trideuteromethyl)propane, 2-Methyl-2-propanol-d10, tert-Butanol-d10, tert-Butanol-D10, tert-Butyl alcohol-d10, 2-(Methyl-d3)-2-propan-1,1,1,3,3,3-d6-ol-d
InChI formel InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3,3D3,5D
SMIL [2H]OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-deuteriooxy-2-(trideuteriomethyl)propane
Molekylvægt (g/mol) 84.183
Formel vægt 84.136