Få mere at vide
tert-Butyl Alcohol-d10, TRC
Artikelnummer. 30427196
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30427196 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427196 Leverandør Toronto Research Chemicals Leverandørnr. TRCT1175411G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butyl Alcohol-d10
CAS 53001-22-2
Molekylær formel C4 D10 O
Mængde 1 g
Synonym 2-Propan-1,1,1,3,3,3-d6-ol-d','2-(methyl-d3)- (9CI), tert-Butyl-d9 alcohol-d (6CI','7CI), 1,1,1,3,3,3-Hexadeutero-2-deuterooxy-2-(trideuteromethyl)propane, 2-Methyl-2-propanol-d10, tert-Butanol-d10, tert-Butanol-D10, tert-Butyl alcohol-d10, 2-(Methyl-d3)-2-propan-1,1,1,3,3,3-d6-ol-d
InChI formel InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3,3D3,5D
SMIL [2H]OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-deuteriooxy-2-(trideuteriomethyl)propane
Molekylvægt (g/mol) 84.183
Formel vægt 84.136