CAS RN 528-76-7
IUPAC Navn:
[(2,4-dinitrophenyl)sulfanyl]chlorane
Synonymer:
[(2,4-dinitrophenyl)sulfanyl]chlorane
Molekylvægt (g/mol):
234.61
Molekylformel:
C6H3ClN2O4S
InChi Key:
GPXDNWQSQHFKRB-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(SCl)C(=C1)[N+]([O-])=O
1
–
3
af
3
Resultater
1
2,4-Dinitrobenzenesulfenyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4-Dinitrobenzenesulfenyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4-Dinitrobenzenesulfenyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.