CAS RN 5202-86-8
IUPAC Navn:
N-(4-chloro-2-methylphenyl)acetamide
Synonymer:
N-(4-chloro-2-methylphenyl)acetamide
Molekylvægt (g/mol):
183.64
Molekylformel:
C9H10ClNO
InChi Key:
WMHHXYOPEQARIN-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C(Cl)C=C1C
1
–
4
af
4
Resultater
1
N-(4-Chloro-2-methylphenyl)acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(4-Chloro-2-methylphenyl)acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(4-Chloro-2-methylphenyl)acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.