CAS RN 51630-58-1
IUPAC Navn:
cyano(3-phenoxyphenyl)methyl 2-(4-chlorophenyl)-3-methylbutanoate
Synonymer:
tirade pydrin, (R-(R*,S*))- aqmatrine epa shaughnessy code: 109301 pydrin, (S-(R*,S*))- (s-(r*,s*))-pydrin sumibac (S-(R*,S*))-isomer pydrin evercide 2362 sumifly
Molekylvægt (g/mol):
419.91
Molekylformel:
C25H22ClNO3
InChi Key:
NYPJDWWKZLNGGM-UHFFFAOYNA-N
SMILES:
CC(C)C(C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1)C1=CC=C(Cl)C=C1
1
–
9
af
9
Resultater
1
Fenvalerate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fenvalerate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fenvalerate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.