Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fenvalerate |
| CAS | 51630-58-1 |
| CAS Max % | ≤ 100% |
| Tæthed | 1.260 g/cm3 (20°C) |
| Beilstein | 2025982 |
| Farve | Light Yellow |
| Smeltepunkt | 39.5°C |
| Synonym | a-Cyano-3-phenoxybenzyl a-(4-chlorophenyl)isovalerate; (αRS)-α-cyano-3-phenoxybenzyl (2RS)-2-(4-chlorophenyl)-3-methylbutyrate |
| Molekylvægt (g/mol) | 419.9 g/mol |
| Mængde | 100 mg |