CAS RN 5154-02-9
IUPAC Navn:
5-hydroxy-1,2-dihydroisoquinolin-1-one
Synonymer:
5-hydroxy-1(2H)-isoquinolinone
Molekylvægt (g/mol):
161.16
Molekylformel:
C9H7NO2
InChi Key:
LFUJIPVWTMGYDG-UHFFFAOYSA-N
SMILES:
OC1=C2C=CNC(=O)C2=CC=C1
1
–
2
af
2
Resultater
1
1,5-Isoquinolinediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,5-Isoquinolinediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.