CAS RN 510-15-6
IUPAC Navn:
ethyl 2,2-bis(4-chlorophenyl)-2-hydroxyacetate
Synonymer:
Acar caswell no. 434 ethyl p,p'-dichlorobenzilate ethyldichlorobenzilate epa pesticide chemical code 028801 chlorbenzylate chlorobenzilate chlorbenzilat geigy 338 kop-mite
Molekylvægt (g/mol):
325.19
Molekylformel:
C16H14Cl2O3
InChi Key:
RAPBNVDSDCTNRC-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(O)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
1
–
5
af
5
Resultater
1
Chlorobenzilate 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality