CAS RN 50910-53-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Ambroxol EP Impurity C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
trans-4-[[(2-Amino-3,5-dibromophenyl)methylene]amino]cyclohexanol (Ambroxol Impurity C), TRC
CAS: 50910-53-7 Molekylær formel: C13 H16 Br2 N2 O Molekylvægt (g/mol): 376.09 Synonym: trans-4-[[(E)-2-Amino-3,5-dibromobenzyliden]amino]cyclohexanol,Ambroxol Hydrochloride Imp. C (EP) IUPAC navn: 4-[(E)-(2-amino-3,5-dibromophenyl)methylideneamino]cyclohexan-1-ol SMIL: Nc1c(Br)cc(Br)cc1\C=N\[C@@H]2CC[C@@H](O)CC2