Få mere at vide
trans-4-[[(2-Amino-3,5-dibromophenyl)methylene]amino]cyclohexanol (Ambroxol Impurity C), TRC
Artikelnummer. 30132432
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132432 500 mg
30007861 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132432 Leverandør Toronto Research Chemicals Leverandørnr. TRCA604605500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50910-53-7
Molekylær formel C13 H16 Br2 N2 O
Molekylvægt (g/mol) 376.09
Synonym trans-4-[[(E)-2-Amino-3,5-dibromobenzyliden]amino]cyclohexanol, Ambroxol Hydrochloride Imp. C (EP)
IUPAC navn 4-[(E)-(2-amino-3,5-dibromophenyl)methylideneamino]cyclohexan-1-ol
SMIL Nc1c(Br)cc(Br)cc1\C=N\[C@@H]2CC[C@@H](O)CC2
Tekniske data
Kemisk navn eller materiale trans-4-[((E)-2-Amino-3,5-dibromobenzyliden)amino]cyclohexanol
Analyt- eller komponentnavne trans-4-[((E)-2-Amino-3,5-dibromobenzyliden)amino]cyclohexanol
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C13H16Br2N2O/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10/h5-7,10-11,18H,1-4,16H2/b17-7+/t10-,11-
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 500 mg
Formel vægt 373.9629
Procent renhed >95%