Få mere at vide
Ambroxol EP Impurity C, Mikromol™
Artikelnummer. 30457885
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457885 4 x 25 mg
30052000 25 mg
17900956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30457885 Leverandør Mikromol Leverandørnr. MM0005.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ambroxol EP Impurity C, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-4-[((E)-2-Amino-3,5-dibromobenzyliden)amino]cyclohexanol
CAS 50910-53-7
Analyt- eller komponentnavne trans-4-[((E)-2-Amino-3,5-dibromobenzyliden)amino]cyclohexanol
Forsendelsestilstand Room Temperature
Molekylær formel C13 H16 Br2 N2 O
Certificeringer/compliance ISO 17025
Synonym trans-4-[[(E)-2-Amino-3,5-dibromobenzyliden]amino]cyclohexanol, Ambroxol Hydrochloride Imp. B (EP), Ambroxol Imp. B (EP), trans-4-[[(E)-2-Amino-3,5-dibromobenzyliden]amino]cyclohexanol, Ambroxol Hydrochloride Imp. C (EP), Ambroxol Hydrochloride Impurity C, Ambroxol Impurity C
InChI formel InChI=1S/C13H16Br2N2O/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10/h5-7,10-11,18H,1-4,16H2/b17-7+/t10-,11-
Løsningstype Neat
SMIL Nc1c(Br)cc(Br)cc1\C=N\[C@@H]2CC[C@@H](O)CC2
IUPAC navn 4-[(E)-(2-amino-3,5-dibromophenyl)methylideneamino]cyclohexan-1-ol
Molekylvægt (g/mol) 376.09
Mængde 4 x 25 mg
Formel vægt 373.9629 g/mol