CAS RN 5015-38-3
IUPAC Navn:
barium(2+) 3-(phosphonatooxy)propanenitrile
Synonymer:
barium(2+) 3-(phosphonatooxy)propanenitrile
Molekylvægt (g/mol):
286.37
Molekylformel:
C3H4BaNO4P
InChi Key:
MRQIDZJGQMWVQR-UHFFFAOYSA-L
SMILES:
[Ba++].[O-]P([O-])(=O)OCCC#N
1
–
2
af
2
Resultater
1
2-Cyanoethyl Phosphate, Barium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Cyanoethyl Phosphate, Barium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.