CAS RN 4994-16-5
IUPAC Navn:
(cyclohex-3-en-1-yl)benzene
Synonymer:
4-phenylcyclohexene
Molekylvægt (g/mol):
158.24
Molekylformel:
C12H14
InChi Key:
XWCWNUSFQVJNDI-UHFFFAOYNA-N
SMILES:
C1CC(CC=C1)C1=CC=CC=C1
1
–
2
af
2
Resultater
1
4-Phenyl-1-cyclohexene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Phenyl-1-cyclohexene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.