CAS RN 497-06-3
IUPAC Navn:
but-3-ene-1,2-diol
Synonymer:
3-butene-1,2-diol
Molekylvægt (g/mol):
88.11
Molekylformel:
C4H8O2
InChi Key:
ITMIAZBRRZANGB-UHFFFAOYNA-N
SMILES:
OCC(O)C=C
1
–
1
af
1
Resultater
1
1-Butene-3,4-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.