CAS RN 474668-76-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(1R,3R)-1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carbo, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.