CAS RN 456-19-9
IUPAC Navn:
1-(dichloromethyl)-4-fluorobenzene
Synonymer:
1-(dichloromethyl)-4-fluorobenzene
Molekylvægt (g/mol):
179.02
Molekylformel:
C7H5Cl2F
InChi Key:
MFIOEEWGBMJNGG-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C(Cl)Cl
1
–
3
af
3
Resultater
1
4-Fluorobenzal Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Fluorobenzal Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Fluorobenzal Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.